Cover for Precision™ Water Baths
Cover for Precision™ Water Baths
Actual product may vary
Cover for Precision™ Water Baths
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
TSPTSGPACL02Clear Polycarbonate Gable CoverPrecision 2L General Purpose Water Bath
TSGPACL02Clear Polycarbonate Gable CoverPrecision 2L General Purpose Water Bath
Catalog number TSPTSGPACL02
Price (SGD)
328.00
EA
Add to cart
Description:
Clear Polycarbonate Gable Cover
For Use With (Equipment):
Precision 2L General Purpose Water Bath
Price (SGD)
328.00
EA
Add to cart
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Gable Covers
  • Enhanced hinged design supports open lid configuration and easy lid removal
  • Accommodate taller labware with extended height
  • Polycarbonate and stainless steel models available
  • Warranty varies by country, please contact us for details
Specifications
DescriptionClear Polycarbonate Gable Cover
Capacity (Metric)2 L
For Use With (Equipment)Precision 2L General Purpose Water Bath
TypeGable Cover
MaterialPolycarbonate
Unit SizeEA