Cover for Precision™ Water Baths
Actual product may vary
Thermo Scientific
Cover for Precision™ Water Baths
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterialFor Use With (Equipment)
TSGPACL02Gable CoverPolycarbonatePrecision 2L Water Bath
TSPTSGPACL02Gable CoverPolycarbonatePrecision 2L Water Bath
Catalog number TSGPACL02
HSN 90278990
Price (SGD)
328.00
Each
Type:
Gable Cover
Material:
Polycarbonate
For Use With (Equipment):
Precision 2L Water Bath
Price (SGD)
328.00
Each
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Gable Covers
  • Enhanced hinged design supports open lid configuration and easy lid removal
  • Accommodate taller labware with extended height
  • Polycarbonate and stainless steel models available
  • Warranty varies by country, please contact us for details
Specifications
Capacity (Metric)2 L
For Use With (Equipment)Precision 2L Water Bath
TypeGable Cover
MaterialPolycarbonate
Unit SizeEach