Search Fisher Scientific
| Catalog Number | Model | Capacity (Metric) |
|---|---|---|
| TSGP28 | Precision GP 28 | 28 L |
| FKS15395837 | Precision GP 02 | 2 L |
| TSGP02 | Precision GP 02 | 2 L |
| TSPTSGP02 | Precision GP 02 | 2 L |
| TSGP05 | Precision GP 05 | 5 L |
| TSPTSGP05 | Precision GP 05 | 5 L |
| TSGP10 | Precision GP 10 | 10 L |
| TSPTSGP10 | Precision GP 10 | 10 L |
| TSGP20 | Precision GP 20 | 20 L |
| TSPTSGP20 | Precision GP 20 | 20 L |
| TSPTSGP28 | Precision GP 28 | 28 L |
| TSGP15D | Precision GP 15D | 15 L |
| TSPTSGP15D | Precision GP 15D | 15 L |