TSG Series Solid Door Laboratory Refrigerators
Actual product may vary
Thermo Scientific
TSG Series Solid Door Laboratory Refrigerators
The TSG Solid Door Refrigerators offer excellent performance for sample storage needing 2-8 ° C, an outstanding 4-year parts and labor warranty, and are available in single and double door configurations. All TSG Series Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigeration.
Have Questions?
Catalog NumberCapacity (English)InteriorElectrical Requirements
TSG5005SV51.1 cu. ft.White (Painted)230V/50Hz
Catalog number TSG5005SV
HSN 84185000
Price (SGD)
9,666.00
Each
Capacity (English):
51.1 cu. ft.
Interior:
White (Painted)
Electrical Requirements:
230V/50Hz
Price (SGD)
9,666.00
Each
Thermo Scientific TSG Series Solid Door Laboratory Refrigerators offer quiet, intuitive operation with excellent temperature performance and an outstanding warranty, meeting the needs of a wide variety of professional users who need consistent, reliable performance without sacrificing usability or sustainability. All TSG Series Laboratory Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigerants and are configured to provide excellent temperature performance while minimizing power consumption, all delivered from our zero waste to landfill facility in Asheville, North Carolina.
The drive for sample integrity and security
  • Designed to safely store media, reagents, and other laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance
Enabling compliance
  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863
Designed for an ergonomic and comfortable experience
  • Ergonomically designed door handle
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned solid doors with 90° stop to assist with inventory loads
Verified sustainability
  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility
Ready for connectivity and monitoring
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
Make it your own
  • Extra and Alternate wire baskets, shelves, and drawers
  • Stainless Steel interior finish available
  • E-Lock Adapter kits
Specifications
DescriptionTSG Series General Purpose Refrigerator
TypeGeneral Purpose Refrigerator
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Temperature Range3°C to 7°C
Temperature (Setpoint)+5°C
RefrigerantR290
Door StyleSolid
No. of Doors2
Shelves8 Adjustable
Capacity (English) Shelf100 lb.
Capacity (Metric) Shelf45 kg
Cabinet MaterialPainted Steel
Exterior FinishPainted
InteriorWhite (Painted)
Certifications/ComplianceCE
StyleUpright
Control TypeCapacitive Touch
Ports1 Rear Access Port
Warranty4 Years Parts and Labor + 1 Year Compressor
Monitoring OptionsSmartVue Pro, Remote Alarm Contacts
Access SecurityKey Lock
CompressorHermetic
Dimensions (DxWxH) Exterior37.9 x 56.5 x 78.5 in. (962 x 1435 x 1994 mm)
Dimensions (D x W x H) Interior28.5 x 52.5 x 58.0 in. (724 x 1334 x 1473 mm)
Plug TypeCEE 7/7
Voltage208/230 V
Electrical Requirements230V/50Hz
Shipping Weight (English)621 lb.
Shipping Weight (Metric)282 kg
Unit SizeEach