Specifications
TypeUpright
Capacity (English)20 cu.ft.
Capacity (Metric)564 L
Temperature Range-15°C to -25°C
Temperature (Setpoint)-20°C
Door StyleSolid
No. of Doors1
Cabinet MaterialPainted Steel
Chamber MaterialPainted Steel
InsulationHFC-free Polyurethane Foam
Exterior FinishWhite-coated Steel
Shelves4
Certifications/ComplianceCE
StyleUpright
DisplayDigital
CastersLocking
Control TypeDigital
Ports1 Rear-access Port
Warranty2 Years Parts & Labor
DefrostAuto
LightingLED
Dimensions (DxWxH) Exterior34.7 x 28.1 x 79.7 in. (88.1 x 71.3 x 202.6 cm)
Dimensions (D x W x H) Interior27.6 x 23.5 x 59.3 in. (70.0 x 59.6 x 150.5 cm)
Monitoring OptionsAudible and Visual High and Low Temperature Alarms
Access SecurityKey Lock
Plug TypeEU
Voltage230 V
Frequency50 Hz
Energy Usage4.42 kw-hr/day
CompressorHermetic
RefrigerantR290
Shipping Weight (English)335 lb.
Shipping Weight (Metric)152 kg
Unit SizeEach