TSE Series Solid Door Laboratory Freezers
Actual product may vary
TSE Series Solid Door Laboratory Freezers
TSE Series Solid Door Laboratory Freezers
TSE Series Solid Door Laboratory Freezers
TSE Series Solid Door Laboratory Freezers
TSE Series Solid Door Laboratory Freezers
TSE Series Solid Door Laboratory Freezers
Thermo Scientific TSE Series Laboratory Freezers deliver dependable cold storage for routine lab needs. Preset to -20°C, these auto-defrost units offer digital control and temperature alarms for easy, reliable use in practical undercounter and upright single door sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Capacity (English)Electrical RequirementsStyle
TSE1220FV266 L9.4 cu. ft.230 V/50 HzUpright
TSE0420FV77 L3 cu.ft.230 V/50 HzUndercounter
TSE2220FV564 L20 cu.ft.230 V/50 HzUpright
Catalog number TSE1220FV
HSN 84184090
Price (SGD)
4,778.00
Each
Capacity (Metric):
266 L
Capacity (English):
9.4 cu. ft.
Electrical Requirements:
230 V/50 Hz
Style:
Upright
Price (SGD)
4,778.00
Each
Thermo Scientific TSE Series Laboratory Freezers provide dependable temperature performance and straightforward operation for routine lab use. Auto-defrost functionality, digital control, and essential features support everyday cold storage needs. All models use SNAP and F-Gas compliant hydrocarbon refrigerants and include a two-year warranty, offering practical value for daily laboratory workflows.
  • Designed for everyday research and academic lab applications with good performance and easy-to-use digital controls
  • Quiet operation, less than 53 dB
  • SNAP compliant green refrigerants
  • Auto defrost
  • Four 2 in. casters, front casters are lockable
  • Hi/Lo Temperature, and door ajar failure visual alarms
  • Rear access port
  • Keyed door locks, compatible with popular E-lock adapters
Stacking Kit accessory is available
Specifications
Capacity (English)9.4 cu. ft.
Capacity (Metric)266 L
Temperature Range-15°C to -25°C
Temperature (Setpoint)-20°C
Door StyleSolid
No. of Doors1
Cabinet MaterialScratch-resistant Painted Steel
Chamber MaterialPainted Steel
InsulationHFC-free Polyurethane Foam
Shelves5
Certifications/ComplianceCE
StyleUpright
DisplayDigital
CastersLocking
Control TypeDigital
Ports1 Rear-access Port
Warranty2 Years Parts and Labor
DefrostAuto
Dimensions (DxWxH) Exterior25.4 x 23.4 x 74.8 in. (64.5 x 59.5 x 190 cm)
Dimensions (D x W x H) Interior19.0 x 19.3 x 62.8 in. (48.3 x 49 x 159.5 cm)
Monitoring OptionsAlarm
Access SecurityKey Lock
Plug TypeUK
Electrical Requirements230 V/50 Hz
Energy Usage1.18 kw-hr/day
CompressorHermetic
RefrigerantR600a
Shipping Weight (English)194 lb.
Shipping Weight (Metric)88 kg
Unit SizeEach