Acetonitrile (ACN), LC-MS Grade
Acetonitrile (ACN), LC-MS Grade
Actual product may vary
Acetonitrile (ACN), LC-MS Grade
An LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.
Have Questions?
Catalog NumberQuantity
TS-51101
also known as PIERCE #51101
1 L
Catalog number TS-51101
also known as PIERCE #51101
Price (SGD)
-
Quantity:
1 L
Thermo Scientific™ Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.
Thermo Scientific LC/MS Grade Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. Acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmosphere with PTFE-lined fluorocarbon caps for ultimate protection.
Highlights:
  • 37 quality tests ensure low, stable baselines and lot-to-lot consistency
  • Low UV absorbance yields low baselines and high detection sensitivity
  • Low impurities protects columns and simplifies analysis by eliminating extraneous peaks
  • Specially purified and tested to the highest specifications to ensure data integrity, maximize assay sensitivity, and to prolong equipment life
  • Store between 2°C to 25°C in a dry, cool, well-ventilated area, away from incompatible materials
Product Details:
  • Alternative names: Methyl cyanide, cyanomethane, ethanenitrile
  • Molecular formula: CH3CN
  • Molecular weight: 41.05
  • Density: 0.780 g/mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
GradeLC-MS
Item NameAcetonitrile (ACN)
Quantity1 L
FormLiquid
Reagent TypeSolvent (Pure)
Product LinePierce™
For Use WithHPLC, Mass Spectrometry
CAS75-05-8
Unit SizeEach
Contents & Storage
Store between 2 to 25°C in a dry, cool, well-ventilated area, away from incompatible materials.
Frequently asked questions (FAQs)
What kind of buffers and solutions are compatible with peptide clean-up and LC-MS?
Most organic solvents or volatile buffers are compatible with LC-MS. Water and acetonitrile with 0.1% formic acid is the most common solvent system for separation of peptide samples for mass spectrometry analysis. Trifluoracetic acid (TFA) is an alternative ion pairing agent for off-line peptide C18 clean-up or LC-UV analysis. We offer the following LC-MS grade reagents: Pierce 0.1% Trifluoroacetic Acid (v/v) in Water (Cat. No. 85172), Pierce Water, LC-MS Grade (Cat. No. 85189), Pierce Acetonitrile (ACN) (Cat. No. 85188), and Pierce Trifluoroacetic (TFA) (Cat. No. 85183).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.