Search Fisher Scientific
| Catalog Number | Description | Volume (Metric) | Tip Style | Filtered |
|---|---|---|---|---|
| TFLR113-100-Q | 100 μL | Beveled | Filtered | |
| TLR102-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered |
| TLR102R-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length, Conical | Non-filtered |
| TLR102RS-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered |
| TLR102RL-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered |
| TLR102RLS-Q | QSP 10XL, Non-Filtered | 10 μL | Extended Length | Non-filtered |
| TFLR113-20-Q | 20 μL | Beveled | Filtered | |
| TLR070-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered |
| TLR070R-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered |
| TLR070RS-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered |
| TLR070RL-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered |
| TLR070RLS-Q | QSP 200, Non-Filtered | 200 μL | Beveled | Non-filtered |
| TLR106-Q | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered |
| TLR106RS-Q | QSP 300, Non-Filtered | 300 μL | MicroPoint | Non-filtered |
| TFLR140-200-Q | 200 μL | MicroPoint | Filtered | |
| TFLR140-300-Q | 300 μL | MicroPoint | Filtered | |
| TLR112NXL-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered |
| TLR112NXLR-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered |
| TLR112NXLRS-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered |
| TLR112NXLRL-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered |
| TLR112NXRLS-Q | QSP 1000, Non-Filtered | 1000 μL | Extended Length | Non-filtered |
| TFLR1121000-Q | 1000 μL | Extended Length | Filtered | |
| TFLR102-10-Q | 10 μL | Extended Length | Filtered |