QSP Low Retention Filtered And Non-Filtered Pipette Tips
Actual product may vary
QSP Low Retention Filtered And Non-Filtered Pipette Tips
Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Catalog NumberDescriptionVolume (Metric)Tip StyleFiltered
TFLR1121000-Q1000 μLExtended LengthFiltered
Catalog number TFLR1121000-Q
HSN 39269099
Price (SGD)
72.00
Special offer
Ends: 30-Jun-2026
89.00
Save 17.00 (19%)
PK
Volume (Metric):
1000 μL
Tip Style:
Extended Length
Filtered:
Filtered
Price (SGD)
72.00
Special offer
Ends: 30-Jun-2026
89.00
Save 17.00 (19%)
PK
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
Featuring:
  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
Volume (Metric)1000 μL
FilteredFiltered
SterilitySterile
ColorNatural
ModelQSP 1000, Filtered
Length (Metric)8.9 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
FormatHinged Rack
Tip StyleExtended Length
For Use With (Equipment)Universal Fit
Product LineQSP
Unit SizePK