▸
Racks
▸
Freezer Storage Racks
▸
Comfort Rack for Upright Freezers, 50 mm Box Height
Actual product may vary
Tenak
Comfort Rack for Upright Freezers, 50 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Have Questions?
Catalog Number
Holds
TENTE24236
Boxes (24)
Catalog number
TENTE24236
Price (SGD)
-
Holds:
Boxes (24)
In the comfort rack, the cryoboxes are placed in drawers
Increases the flexibility and reduces the operating weight compared to the rack types
Measure the inside height and depth of you freezer when choosing the right rack
Racks can be used with all freezer models
Constructed of stainless steel
Specifications
Height (Metric)
50 mm (Box)
Dimensions (L x W x H)
562 x 140 x 340 mm
For Use With (Equipment)
Upright Freezers
Holds
Boxes (24)
Array
6 x 4
Material
Stainless Steel