Origami Cardboard Cryobox
Actual product may vary
Fisherbrand
Origami Cardboard Cryobox
Save space in the laboratory with Tenak™Origami Cardboard Cryobox. The box consists of a lid and a bottom with instructions to create a stable, high quality square cryobox..
Have Questions?
Catalog NumberHeight (Metric)Includes
TENTE2282550 mm9 x 9 Divider
Catalog number TENTE22825
Price (SGD)
-
Height (Metric):
50 mm
Includes:
9 x 9 Divider
  • Spacesaving in the laboratory
  • Square measure of 134x134mm fits all storage rack systems
  • Packages of 100 each
.
Specifications
MaterialCardboard
ShapeSquare
TypeCryo Box
Includes9 x 9 Divider
Quantity100 Pack
Width (Metric)134 mm
Height (Metric)50 mm
Weight (Metric)7 kg
Unit SizePack