▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
30 mm Cryobox Dividers
Actual product may vary
Tenak
30 mm Cryobox Dividers
Dividers for use with 50 mm high cryoboxes.
Have Questions?
Change view
Catalog Number
Holds
Tube Array
TENTE22688
Vials with 25.25 mm O.D.
5 x 5 cells
TENTE22113
Vials with 24 mm O.D.
5 x 5 cells
2 Options
Catalog number
TENTE22688
Price (SGD)
-
Holds:
Vials with 25.25 mm O.D.
Tube Array:
5 x 5 cells
For use in cardboard cryoboxes
Holds cryogenic vials
Grid style divider
Specifications
Type
Grid Divider
For Use With (Equipment)
50 mm high cryoboxes
Holds
Vials with 25.25 mm O.D.
Tube Array
5 x 5 cells
Height (Metric)
30 mm