▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
Polypropylene Cryoboxes
Actual product may vary
Tenak
Polypropylene Cryoboxes
Have Questions?
Catalog Number
Color
Holds
TENTE22300
Blue
1.2 to 2 mL Cryogenic Vials
Catalog number
TENTE22300
Price (SGD)
-
Color:
Blue
Holds:
1.2 to 2 mL Cryogenic Vials
Specifications
Color
Blue
Material
Polypropylene
Shape
Square
Type
Cryo Box
Holds
1.2 to 2 mL Cryogenic Vials
Quantity
36 Pack
Length (Metric)
130 mm
Width (Metric)
130 mm
Height (Metric)
50 mm
Weight (Metric)
0.08 kg