▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
Grid Dividers
Actual product may vary
Tenak
Grid Dividers
Organize and retrieve samples stored in Cryoboxes into which Tenak™ Grid Dividers have been inserted. Boxes and grids hold tissue in a variety of sample storage tube sizes and diameters.
Have Questions?
Change view
Catalog Number
Holds
Height (Metric)
TENTE22150
6 Vials (Less than 20 mm O.D.)
65 mm
TENTE22110
9 Vials (Less than 13 mm O.D.)
30 mm
TENTE22127
6 Vials (Less than 20 mm O.D.)
40 mm
3 Options
Catalog number
TENTE22150
Price (SGD)
-
Holds:
6 Vials (Less than 20 mm O.D.)
Height (Metric):
65 mm
Ultra-low temperature-resistant cardboard
Water-repellent finish
Temperature resistance to -80°C
Specifications
Material
Cardboard
Shape
Square
Type
Grid Divider
For Use With (Application)
For 100 and 130mm Boxes
For Use With (Equipment)
100 and 130 mm Boxes
Holds
6 Vials (Less than 20 mm O.D.)
Quantity
36 Pack
Height (Metric)
65 mm