65 mm Cryobox Dividers
Actual product may vary
Tenak
65 mm Cryobox Dividers
Dividers for use with 100 and 130 mm high cryoboxes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsTube Array
TENTE22148Vials with 31 mm O.D.4 x 4 cells
FKS15240554Vials with 31 mm O.D.4 x 4 cells
FKS15250554Vials with 24 mm O.D.5 x 5 cells
TENTE22149Vials with 24 mm O.D.5 x 5 cells
TENTE22142Vials with 15 mm O.D.8 x 8 cells
TENTE22144Vials with 13 mm O.D.9 x 9 cells
Catalog number TENTE22148
Price (SGD)
-
Holds:
Vials with 31 mm O.D.
Tube Array:
4 x 4 cells
  • For use in cardboard cryoboxes
  • Holds cryogenic vials
  • Grid style divider
Specifications
TypeGrid Divider
For Use With (Equipment)100 and 130 mm high cryoboxes
HoldsVials with 31 mm O.D.
Tube Array4 x 4 cells
Height (Metric)65 mm
Unit SizeCase