▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
Grid Divider for Cardboard Cryoboxes 133 x 133mm
Actual product may vary
Tenak
Grid Divider for Cardboard Cryoboxes 133 x 133mm
For safe and secure storage in the laboratory. Tenak™ Cardboard Cryoboxes 133 x 133mm are made of fibreboard/cardboard.
Have Questions?
Catalog Number
Holds
TENTE22140
49 Vials
Catalog number
TENTE22140
Price (SGD)
160.00
Pack
Add to cart
Holds:
49 Vials
Price (SGD)
160.00
Pack
Add to cart
White in color
Water-repellent for longer durability
Divider is also available
Specifications
Description
Grid divider
Material
Cardboard
Shape
Square
Type
Grid Divider
Holds
49 Vials
Tube Array
7 x 7
Height (Metric)
65 mm