▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
40 mm Cryobox Dividers
Actual product may vary
Tenak
40 mm Cryobox Dividers
Dividers for use with 75 mm high cryoboxes.
Have Questions?
Change view
Catalog Number
Holds
Tube Array
TENTE22134
Vials with 13 mm O.D.
9 x 9 cells
TENTE22130
Vials with 17 mm O.D.
7 x 7 cells
2 Options
Catalog number
TENTE22134
Price (SGD)
137.00
Pack
Add to cart
Holds:
Vials with 13 mm O.D.
Tube Array:
9 x 9 cells
Price (SGD)
137.00
Pack
Add to cart
For use in cardboard cryoboxes
Holds cryogenic vials
Grid style divider
Specifications
Type
Grid Divider
For Use With (Equipment)
75 mm high cryoboxes
Holds
Vials with 13 mm O.D.
Tube Array
9 x 9 cells
Height (Metric)
40 mm