40 mm Cryobox Dividers
Actual product may vary
Tenak
40 mm Cryobox Dividers
Dividers for use with 75 mm high cryoboxes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsTube Array
TENTE22130Vials with 17 mm O.D.7 x 7 cells
TENTE22134Vials with 13 mm O.D.9 x 9 cells
Catalog number TENTE22130
Price (SGD)
137.00
Pack
Holds:
Vials with 17 mm O.D.
Tube Array:
7 x 7 cells
Price (SGD)
137.00
Pack
  • For use in cardboard cryoboxes
  • Holds cryogenic vials
  • Grid style divider
Specifications
TypeGrid Divider
For Use With (Equipment)75 mm high cryoboxes
HoldsVials with 17 mm O.D.
Tube Array7 x 7 cells
Height (Metric)40 mm
Unit SizePack