Grid Dividers
Actual product may vary
Tenak
Grid Dividers
Organize and retrieve samples stored in Cryoboxes into which Tenak™ Grid Dividers have been inserted. Boxes and grids hold tissue in a variety of sample storage tube sizes and diameters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsHeight (Metric)
TENTE221109 Vials (Less than 13 mm O.D.)30 mm
TENTE221506 Vials (Less than 20 mm O.D.)65 mm
TENTE221276 Vials (Less than 20 mm O.D.)40 mm
Catalog number TENTE22110
Price (SGD)
87.00
Pack
Holds:
9 Vials (Less than 13 mm O.D.)
Height (Metric):
30 mm
Price (SGD)
87.00
Pack
  • Ultra-low temperature-resistant cardboard
  • Water-repellent finish
  • Temperature resistance to -80°C
Specifications
MaterialCardboard
ShapeSquare
TypeGrid Divider
For Use With (Application)For 50mm Boxes
For Use With (Equipment)50 mm Boxes
Holds9 Vials (Less than 13 mm O.D.)
Quantity36 Pack
Height (Metric)30 mm
Unit SizePack