Check Order Status
Cardboard Cryoboxes 133 x 133mm
Actual product may vary
Cardboard Cryoboxes 133 x 133mm
For safe and secure storage in the laboratory. Tenak™ Cardboard Cryoboxes 133 x 133mm are made of fibreboard/cardboard.
Have Questions?
Catalog number
TENTE22080
Price (SGD)
291.00
PK
Add to cart
Price (SGD)
291.00
PK
Add to cart
White in color
Water-repellent for longer durability
Divider is also available
Specifications
Description
Cryobox
Color
White
Material
Cardboard
Shape
Square
Type
Cryo Box
Dimensions (L x W x H)
133 x 133 x 130 mm
Length (Metric)
133 mm
Width (Metric)
133 mm
Height (Metric)
130 mm
Weight (Metric)
0.11 kg