▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
Red Cardboard Cryobox
Actual product may vary
Tenak
Red Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Have Questions?
Catalog Number
Dimensions (L x W x H)
TENTE22060
133 x 133 x 75 mm
Catalog number
TENTE22060
Price (SGD)
219.00
Pack
Add to cart
Dimensions (L x W x H):
133 x 133 x 75 mm
Price (SGD)
219.00
Pack
Add to cart
Constructed of fibreboard/cardboard
Water-repellent for longer durability
Can be used with a large range of grid dividers
Specifications
Color
Red
Material
Cardboard
Shape
Square
Type
Cryo Box
Dimensions (L x W x H)
133 x 133 x 75 mm