TDE Series -86°C Ultra-Low Temperature Freezer,
Actual product may vary
Thermo Scientific
TDE Series -86°C Ultra-Low Temperature Freezer,
Part of the Standard Performance (STP) family, Thermo Scientific™ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models, maximizing storage capacity from 30,000 up to 60,000 2 mL vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical RequirementsCapacity (Metric)Certifications/Compliance
TDE50086FV-ULTS
also known as 25CVT4E01A
230V, 50 Hz, 1 Phase682 LCE, cUL, cUL, ISO 13485
TDE40086FV-ULTS
also known as 24CVT4E01A
230V, 50 Hz, 1 Phase549 LCE
TDE60086FV-ULTS
also known as 26CVT4E01A
230V, 50 Hz, 1 Phase816 LCE, cUL, UL, ISO 13485
Catalog number TDE50086FV-ULTS
also known as 25CVT4E01A
HSN 84184090
Price (SGD)
17,969.00
Each
Electrical Requirements:
230V, 50 Hz, 1 Phase
Capacity (Metric):
682 L
Certifications/Compliance:
CE, cUL, cUL, ISO 13485
Price (SGD)
17,969.00
Each
Part of the Standard Performance (STP) family, Thermo Scientific™ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models, maximizing storage capacity from 30,000 up to 60,000 2 mL vials.
The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab. From our first ULT freezer in 1940 to today, the combined experience of REVCO, Forma Scientific, Jewett, HERAFreeze, and other families of products continues to drive our ability to deliver industry-leading innovations that help accelerate science and health care.
Performance
  • Powered by H-drive's hydrocarbon technology, these systems save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers1
  • Designed for daily usage, recovers cabinet temperature 20% faster after opening the door.2
  • A quiet sound output below 53 dBA, enables the freezer to come out of the hallway and into the lab
  • Meets ±10°C validation protocols at -80°C with strong peak variation and steady-state temperature variation
  • Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection
User Friendly
  • The H-drive Information Center (HIC) combines easy to read temperature with simple capacitive touch buttons in a 5.6 inch display
  • Four capacities (300, 400, 500 and 600 2-inch box) to accommodate a variety of storage needs and lab footprint specifications
  • All models contain four inner doors allowing for easy sample access without exposing greater freezer content to ambient conditions
  • Improved customer convenience with new ergonomically designed handle, high contrast easy-to-read display, and comfortable easy-access shelf locations.
  • Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
  • Security features including alarm settings and key-lock door
Sustainability
  • Lower HVAC costs as a result of less heat emission into the environment
  • Natural refrigerants and Polyurethane foam with non-HFC blowing agent having zero GWP, making the Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), the European Union's F-Gas and other sustainability standards
Get Connected
The Thermo Scientific™ DeviceLink™ Connect remote monitoring solution helps provide peace of mind by enabling you to keep track of your equipment anytime, anywhere. This system supports independent sensors to monitor your instrument, but it can also be connected via serial port directly to the instrument, harnessing the on-board telemetry data, alarms, and events. All information is transmitted to the Thermo Fisher Connect cloud platform where you have 24/7 access to the information via InstrumentConnect's web- and mobile-based applications. For more information visit www.thermofisher.com/dlc.
Compatible DeviceLink Connect System
  • DLCKTULTS101 — Ultra-low temperature freezer
*Each kit includes a DeviceLink Connect system, 1-year basic monitoring subscription, power cable, commissioner USB, mounting hardware, and an applicable serial or USB cable.
Independent DeviceLink Connect Sensors
  • DLCSKTEMP001 — DeviceLink Connect PT100 (-200°C to +600°C) temperature sensor and Smart-Cable adapter
  • DLCSKCURT001 — DeviceLink Connect current transducer (0-50A) with Smart-Cable adapter
  • DLCPTDOOR001 — Door Switch
Footnotes:
  1. Internal data comparing 11.2 kWh/day to UXF600 = 14.9 kWh/day. Data on file.
  2. Typical freezer test data based on internal testing in ambient temperature at 20°C; Typical standard validation protocol of ±10°C.
Specifications
Capacity (English)24.1 cu. ft.
Capacity (Metric)682 L
Temperature Range-50°C to -86°C
Door StyleLatched Pull-out Swing
No. of Doors1 (1 compartment)
Inner Doors4
Door Opening Recovery16 min.
Hinge SideLeft
Cabinet MaterialCold-rolled Steel
Chamber MaterialCold-rolled Steel
InsulationVacuum, Foam
InteriorPainted steel
Exterior FinishPowder-coated Steel
Shelves3
DrawersNone
No. of Racks20
No. of Racks per Kit5
Certifications/ComplianceCE, cUL, cUL, ISO 13485
StyleUpright
DisplayHIC Interface, 7-Segment Display with capacitive touch screen
Casters4
ColorGray
Condenser TypeFan-forced
For Use With (Application)Storage of General (non-flammable) Laboratory Materials
For Use With (Equipment)Storage Racks, Storage Boxes, Vials
IncludesShelves, power plug
PortsYes
Probe TypeSingle RTD (1000 ohm Platinum RTD)
Typical Lifetime10 years
Warranty5 years parts
DefrostManual
Min. Clearance30 cm
Peak Variation at -80C+4.2/-3.8
Warm up Time -80 C to -50C312 minutes
Water CooledNo water cooling
Depth (English) Exterior38.5 in.
Depth (English) Interior28.3 in
Depth (Metric) Exterior97.7 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior1981 mm
Height (Metric) Interior1300 mm
Length (English) Exterior38.5 in.
Length (Metric) Exterior978 mm
Length (English) Interior28.3 in.
Length (Metric) Interior719 mm
Width (English) Exterior34 in.
Width (English) Interior28.8 in.
Width (Metric) Exterior864 mm
Width (Metric) Interior732 mm
Monitoring OptionsAvailable Separately
Access SecurityKey Lock Standard, Padlock Compatible
Chart RecorderOptional
Data OutputsRS485/4-20mA output/Dry Contacts
Operating ModesStandard Mode
Setpoint SecurityYes
Backup SystemsCO2 or LN2 Optional
Smart View CompatibilityYes
Amperage6.5 Amp
Plug TypeEU
Voltage230 V
Electrical Requirements230V, 50 Hz, 1 Phase
Frequency50 Hz
PhaseOne
Energy Usage10.3 kWh/day
Green FeaturesACT Ecolabel (Gold), China Energy Label
Vial Capacity50,000
CompressorH-drive
RefrigerantR290/R170+R290 Mix
Shipping Weight (English)640 lbs.
Shipping Weight (Metric)333 kg
Weight (English)640 lbs.
Weight (Metric)290 kg
Holds50,000 sample vials
Rack Capacity20
Unit SizeEach