2.0mL Graduated, free-standing & attached TC with O-Ring, Sterile
2.0mL Graduated, free-standing & attached TC with O-Ring, Sterile
Actual product may vary
2.0mL Graduated, free-standing & attached TC with O-Ring, Sterile
Available in conical and free standing designs, screw cap microcentrifuge tubes are available in volume capacities of 0.5mL, 1.5 mL and 2.0mL.
Have Questions?
Catalog NumberColor
TC-520-GRDS-QNatural
Catalog number TC-520-GRDS-Q
Price (SGD)
147.00
PK
Add to cart
Color:
Natural
Price (SGD)
147.00
PK
Add to cart
Thermo Scientific™ QSP Screw Cap Microcentrifuge Tubes are available in conical and free standing designs. Natural and amber. These tubes are autoclavable, boil proof and freezable to -80°C. Find a complete volume range from 0.5mL through 2.0 mL.
Screw Cap Microcentrifuge Tube Design
  • Tubes are designed with conical bottoms allowing for more complete sample retrieval
  • Screw caps with O-rings guard against leakage by forming a positive seal against the rim of the tube
  • Tethered screw caps remain attached to the sample tube but can rotate completely for convenience
  • Tubes and caps are available in amber to protect your light sensitive samples
  • External ribs on the tubes allow for single-handed opening and closing when placed in racks with serrated holes
  • Screw cap microcentrifuge Tubes work with many automated systems
  • O-ring is manufactured from durable ethylene polypropylene dimonomere compound
Highest Quality
  • Manufactured from virgin polypropylene these microcentrifuge tubes ensure sample integrity and security during boiling, freezing and centrifugation
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogen
  • Sterile product is certified free of RNase, DNase, DNA, endotoxin, pyrogen and PCR inhibitors
Color/Packaging Options
  • Sterile product is packaged with the caps screwed onto the tubes
  • Nonsterile tubes are packaged in individually sealed bags of 500 to protect against cross contamination
  • Standard Screw Cap Color options are: Blue, Green, Orange, Red, Yellow and Mixed to aid with visual identification of samples
Recommended For: Daily laboratory usage including centrifugation and short term sample storage
Specifications
Cap StyleScrew cap, tethered w/o-ring
Certifications/ComplianceFree of Rnase, Dnase, DNA, Pyrogen,PCR Inhibtor,ATP
DescriptionQSP 2.0mL
AutoclavableAutoclavable
Capacity (Metric)2.0 mL
ColorNatural
MaterialPolypropylene
PackagingBulk
Volume (Metric)2.0mL