Nalgene™ Square PETG Media Bottles with Closure
Nalgene™ Square PETG Media Bottles with Closure
Actual product may vary
Nalgene™ Square PETG Media Bottles with Closure
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure, ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
TBP2019-10001 L
FIS03-311-1V30 mL
FKS1003795030 mL
FIS03-311-1W60 mL
FIS03-312-1125 mL
FIS03-312-3250 mL
FKS10028010250 mL
FIS03-312-4500 mL
FIS03-312-51 L
FIS03-311-1X2000 mL
FKS131386432000 mL
Catalog number TBP2019-1000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1 L
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure. Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates, and also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
Features:
  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Suitable for frozen storage down to -40°C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Sterile and non-pyrogenic
  • Available in low particulate format
  • Supported by extensive validation documentation
Ordering Information:
  • Packed in shrink-wrapped trays
Note:
  • 2 L size has molded-in handgrips
Warranty:
  • 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (Metric)1 L
Capacity (English)32 oz.
ColorClear
Diameter (English)3.7 in.
Diameter (English) Inner Neck1.10 in.
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage
MaterialPETG
No. per Case24/Cs.
No. per Pack12/Pk.
SterilitySterile
Temperature Range (Metric)-70°C to 70°C
Certifications/ComplianceBottles and closures are certified radiation sterilized (SAL 10-6), non-pyrogenic and non-cytotoxic, and comply with USP Class VI guidelines.
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)220 mm
Product TypeMedia Bottle
Height (English)8.67 in.
Closure MaterialHDPE
Closure Size38-430 mm
Diameter (Metric)94 mm
Unit SizeCS