XL Graduated MicroPoint Pipette Tips, 100-1250 μL
Actual product may vary
Thermo Scientific
XL Graduated MicroPoint Pipette Tips, 100-1250 μL
Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorFormatSterility
T112XLRL-QBlueRefillNon-sterile
112NXL-QNaturalBulkNon-sterile
T112XLR-QBlueHinged RackNon-sterile
Catalog number T112XLRL-Q
HSN 84799090
Price (SGD)
81.00
PK
Color:
Blue
Format:
Refill
Sterility:
Non-sterile
Price (SGD)
81.00
PK
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.
Featuring:
  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
DescriptionQSP 1000, Non-Filtered
Volume (Metric)1250 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlue
Length (Metric)8.9 cm
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
FormatRefill
Tip StyleExtended Length
Green FeaturesACT Ecolabel (Silver)
For Use With (Equipment)Universal Fit
GraduatedYes
Product LineQSP
MaterialVirgin Polypropylene
Unit SizePK