Search Thermo Fisher Scientific
Catalog Number | Packaging |
---|---|
T112NXLRL-Q | 4800/Cs. |
QSPT112NXLRL-Q | 960/Pk. |
Thermo Scientific™ QSP™ Graduation Mark Pipette natural color tips 1000μl, micropoint graduated at 100, 200, 500, 1000μl. Extended length 8.89cm. Maximum liquid volume 1250μl. Packaged reload system non-sterile with clean claims; Rnase/Dnase, DNA, pyrogen and endotoxin free. Universal fit on most pipettes.