SteriWare™ HDPE Disposable Powder Thief
SteriWare™ HDPE Disposable Powder Thief
Actual product may vary
SteriWare™ HDPE Disposable Powder Thief
SteriWare™ HDPE Disposable Powder Thief
SteriWare™ HDPE Disposable Powder Thief
SteriWare™ HDPE Disposable Powder Thief
SteriWare™ HDPE Disposable Powder Thief
SteriWare™ HDPE Disposable Powder Thief
Ideal for taking representative samples from known locations within the bulk powder. Powder is trapped within the sampler and easily withdrawn and emptied.
Have Questions?
Catalog NumberCapacity (Metric)Sterility
SPS8060H-510S10 mLSterile
Catalog number SPS8060H-510S
Price (SGD)
-
Capacity (Metric):
10 mL
Sterility:
Sterile
Key Features:
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Available in a variety of lengths and tip sizes
Operation:
  • Insert the sampler into the product ensuring the tip is inside the sampler body.
  • At the required depth pull up the body to expose the tip. The sample will fall in around the tip.
  • Push down body of the sampler to trap the sample.
  • Remove sampler from the product.
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable sampler, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Specifications
Product TypePowder Thief
MaterialHDPE
Handle TypeHDPE
Capacity (Metric)10 mL
DisposableSingle-use
SterilitySterile
Certifications/ComplianceConforms to FDA CFR 177.1520, EU Regulations 10/2011, EC Regulations 1935/2004
For Use With (Application)Ideal for taking representative samples from known locations within the bulk powder
Length (Metric)500 mm
Unit SizePack