SteriWare™ Disposable LiquiThief
SteriWare™ Disposable LiquiThief
Actual product may vary
SteriWare™ Disposable LiquiThief
SteriWare™ Disposable LiquiThief
SteriWare™ Disposable LiquiThief
Ideal for use with low viscocity liquids. Sampling Systems™ SteriWare™ Disposable LiquiThief is essentially a big syringe that pulls the liquid into the sampler. The further the handle is pulled the bigger the sample volume.
Have Questions?
Catalog NumberCapacity (Metric)Sterility
SPS8050H-500S100 mLSterile
Catalog number SPS8050H-500S
Price (SGD)
-
Capacity (Metric):
100 mL
Sterility:
Sterile
Key Features:
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas (500mm and 1000mm lengths only)
  • Available in 3 different sizes
Operation:
  • Insert the sampler into the product.
  • At the required depth pull up the handle. The further the handle is pulled, the larger the sample volume.
  • Withdraw sampler from the product.
  • Push handle down to expel the sample.
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable sampler, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6.
Specifications
Product TypeLiquiThief
MaterialHDPE
Capacity (Metric)100 mL
DisposableSingle-use
SterilitySterile
Diameter (Metric) Outer21 mm
Length (Metric) Shaft500 mm
Unit SizeBox