Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™
Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™
Actual product may vary
Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™
Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesLength (Metric) Cable
SPE8107BNUMDBNC (Waterproof), 8-pin Mini-DIN1 m
FIS13-641-981BNC (Waterproof), 8-pin Mini-DIN1 m
FKS11839221BNC (Waterproof), 8-pin Mini-DIN1 m
FIS13-641-982BNC (Waterproof), 8-pin Mini-DIN3 m
FKS11849221BNC (Waterproof), 8-pin Mini-DIN3 m
Catalog number SPE8107BNUMD
Price (SGD)
-
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
Length (Metric) Cable:
1 m
Thermo Scientific™ Orion™ ROSS Ultra Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra line with a built-in ATC. These products offer a low-maintenance, gel-filled, epoxy body and are best used where minimal maintenance is required. Non-refillable Orion ROSS Ultra Triodes come with an 18 month warranty.
Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonomous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required 
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS
                 pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm
                 electrode storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Warranty period
18 months from date of purchase
Specifications
DescriptionOrion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable
Connector TypesBNC (Waterproof), 8-pin Mini-DIN
JunctionGlass Capillary
MaterialRugged Epoxy Body, Filled with Low Maintenance Gel
pH Range0 to 14
Temperature (Metric) Operating0°C to 80°C
TypepH/ATC Combination Electrode
Filling Solution TypeNone required
For Use With (Equipment)Orion Versa Star, Star A and Dual Star Meters
Internal ReferenceROSS
Isopotential Point7
pH Relative Accuracy0.03 pH for buffers using automatic temperature compensation
Precision0.01
Sample CompatibilityTRIS, sulfides, or proteins
Warranty18 Months
Length (Metric)120 mm
Length (Metric) Cable1 m
Diameter (Metric)12 mm
Unit SizePieces