Isotemp™ Hot Plate Stirrer, Ambient to 540°C, Ceramic
Isotemp™ Hot Plate Stirrer, Ambient to 540°C, Ceramic
Actual product may vary
Isotemp™ Hot Plate Stirrer, Ambient to 540°C, Ceramic
Isotemp™ Hot Plate Stirrer, Ambient to 540°C, Ceramic
Isotemp™ Hot Plate Stirrer, Ambient to 540°C, Ceramic
Isotemp™ Hot Plate Stirrer, Ambient to 540°C, Ceramic
Slow-speed stirring, consistent speed control, and strong magnetic coupling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMax. Container SizeSizeDimensionsLxWxH
SP888542061 L4.25 x 4.25 in.
FKS153635184 L7.25 x 7.25 in.
Catalog number SP88854206
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Max. Container Size:
1 L
Size:
4.25 x 4.25 in.
Experience precise stirring control, exceptional safety and temperature performance for your routine protocols.
Advanced technology
  • Slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
  • Includes stir bar
Specifications
Top Plate MaterialCeramic
Voltage230 V
DescriptionHotplate Stirrer
IncludesStir Bar
Electrical Requirements230 V 50/60 Hz
Length (English)5 in.
Size4.25 x 4.25 in.
Height (Metric)9.1 cm
Temperature (Metric)Ambient to 540°C
Height (English)3.6 in.
Frequency50/60 Hz
Width (Metric)25.4 cm
Length (Metric)12.7 cm
Width (English)10 in.
TypeStirring Hotplate, Stirrer
ShapeSquare
Plug TypeUK/AUS/CN
Max. Container Size1 L
Stirring Range50 to 1500 rpm
Warranty3 Years
Dimensions (LxWxH)10 in. x 5 in. x 3.6 in., 25.4 cm x 12.7 cm x 9.1 cm
Certifications/ComplianceRoHS, cCSAus (100 to 120 V Model), CE (230 V Model)
Unit SizeEA