Optima™ Rubber Gloves
Actual product may vary
Thermo Scientific
Optima™ Rubber Gloves
BM Polyco™ Optima™ Rubber Gloves are made of natural rubber and perform well in protecting against acids, alcohol, and other chemicals.
Have Questions?
Catalog NumberSizeColor
SMO2257, 7.5Pink
Catalog number SMO225
Price (SGD)
-
Size:
7, 7.5
Color:
Pink
  • Medium-weight offering improved strength while maintaining a high level of dexterity
  • Slip-resistant pattern on the palm and fingers ensuring a good level of grip especially in wet conditions
  • Anatomical shape and flexibility minimizes hand fatigue
  • Added surface to provide extra grip during application
  • Chemically resistant
  • Typically used for cleaning and janitorial, manufacturing and assembly, general handling and processing, kitchen work
  • Blue Optima™ gloves are suitable for food handling
Specifications
DescriptionEN 388 Abrasion Resistance Level: 1
ColorPink
MaterialRubber
Product TypeChemically Resistant Natural Rubber Gloves
Size7, 7.5
Thickness (Metric)0.52 mm
For Use With (Application)Typically used for cleaning and janitorial, manufacturing and assembly, general handling and processing, kitchen work.
Liner MaterialCotton Flocking
LiningLined
Length (Metric)300 mm
Unit SizePack