Smart-Vue™ Alert
Product Image
Smart-Vue™ Alert
Replaces the use of an analog USB model for issuing Smart-Vue wireless monitoring alerts via voice and/or SMS.
Have Questions?
Catalog NumberDescriptionRequirementsAlarm Types
SMARTVUEALERT20Smart-Vue Alert, 1-20 Sensors36 month software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors (dual modules count as 2 sensors). Requires SmartVueServer/SmartVue Client version 2.2 and above.Voice, SMS
Catalog number SMARTVUEALERT20
Price (SGD)
3,978.00
EA
Add to cart
Description:
Smart-Vue Alert, 1-20 Sensors
Requirements:
36 month software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors (dual modules count as 2 sensors). Requires SmartVueServer/SmartVue Client version 2.2 and above.
Alarm Types:
Voice, SMS
Price (SGD)
3,978.00
EA
Add to cart
Smart-Vue™ Alert is an online alert platform, replacing the use of an analog USB modem for issuing alerts via voice and/or SMS. This is a requirement in many regions due to the phase-out of analog telephone lines. Smart-Vue Alert is a for-pay service with a yearly subscription model.
Feature summary
  • 12-month or 36-month options, time counting from the date of installation
  • Module-based license options with same ranges as server licenses: 1-20, 21-50, 51-100, 101-unlimited
  • Customer benefits from an unlimited number of alerts/calls for the allowed number of modules
  • Smart-Vue Alert supports both SMS/Text and Voice alerts
  • There is no “text-only” option (i.e. without voice alerts) at this time.
  • Users cannot customize alert messages. This feature could be studied for a future release if necessary.
Specifications
TypeSmart-Vue Alert
Display TypeNo Display
CalibrationNot Calibrated
Alarm TypesVoice, SMS
Software TypeSmart-Vue Client Version 2.2, Smart-Vue Server
DescriptionSmart-Vue Alert, 1-20 Sensors
Requirements36 month software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors (dual modules count as 2 sensors). Requires SmartVueServer/SmartVue Client version 2.2 and above.
For Use With (Application)Wireless Monitoring
Unit SizeEA