MaxQ™ HP Incubated and Refrigerated Console Shakers
MaxQ™ HP Incubated and Refrigerated Console Shakers
Actual product may vary
MaxQ™ HP Incubated and Refrigerated Console Shakers
MaxQ™ HP Incubated and Refrigerated Console Shakers
MaxQ™ HP Incubated and Refrigerated Console Shakers
MaxQ™ HP Incubated and Refrigerated Console Shakers
MaxQ™ HP Incubated and Refrigerated Console Shakers
MaxQ™ HP Incubated and Refrigerated Console Shakers
Perform shaking applications on 4L and 6L flasks with these incubated and refrigerated console shakers,which feature a foot pedal and large viewing window.
Have Questions?
Catalog NumberDescriptionElectrical RequirementsSpeed RangeTemperature Range (Metric)
SHKE481HPIncubated/Refrigerated Floor-Model Console Orbital Shaker230 V, 50 Hz25 to 525 rpm, ±1 rpm4°C to 60°C
Catalog number SHKE481HP
Price (SGD)
43,117.00
EA
Add to cart
Description:
Incubated/Refrigerated Floor-Model Console Orbital Shaker
Electrical Requirements:
230 V, 50 Hz
Speed Range:
25 to 525 rpm, ±1 rpm
Temperature Range (Metric):
4°C to 60°C
Price (SGD)
43,117.00
EA
Add to cart
Utilize Thermo Scientific™ MaxQ™ HP Incubated and Refrigerated Console Shakers for large 4L and 6L flasks. These shakers feature a foot pedal for hands-free operation for loading of samples and a large viewing window for viewing samples without disturbing the chamber atmosphere.
Features:
  • Shakers feature HEPA filtration for maximum protection from contamination
  • Directed airflow system for optimum temperature uniformity and ideal growth conditions
  • Ergonomic design ensures convenient access, and a front drain system makes the chamber easy to clean
Microprocessor Control:
  • Operating temperatures: Incubated: 5°C above ambient to 60°C (41°F to 140°F) or Incubated/Refrigerated: 4°C above ambient to 60°C (39.2° to 140°F) in 0.1°C increments with ±0.1°C control
  • Displays run and setpoints of temperature, speed and time simultaneously on a 20-character backlit display
  • Alphanumeric status messages on cycle status
  • Menu-driven programming for quick, easy access to all parameters
  • Audible/visual over- and undertemperature tracking and safety alarms ensure product protection
  • Audible/visual, high/low rpm tracking alarms alert to speed deviations
Performance:
  • Unique HEPA filtration system
  • HEPA filter rated a minimum 95% efficient at 0.3μm for entrapment of extraneous airborne particulats
  • Laminar airflow across the chamber aids temperature uniformity
  • Low-end temperature achieved by filtered, front-to-back refrigeration system airflow
Construction:
  • Crevice-free, stainless-steel interior with coved corners for easy cleaning
  • Drain system with valve in front of chamber for easy liquid elimination
  • Lid opens by hand or by depressing foot pedal for hands-free operation when loading vessels
  • Angled sides, lower front, and convenient platform height ensure a comfortable reach into chamber
  • Thermal pane window provides a large 2.75 sq. ft. (0.3m2) viewing area
  • Access port (back of cabinet wall) for probes, CO2 gassing manifold, or other equipment
  • Optional shelf provides additional storage
Ordering Information:
Platform not included.
Specifications
Orbit25 mm
Load Bearing Capacity (Metric)32 kg
Speed Range25 to 525 rpm, ±1 rpm
Electrical Requirements230 V, 50 Hz
Voltage230 V
Temperature Range (Metric)4°C to 60°C
Amperage8 A
Holds4 L, 6 L Flask
Product TypeShaker
Load Bearing Capacity (English)71 lb.
Certifications/ComplianceCE, cUL, UL
Plug TypeCEE 7/7 plug
Temperature Range (English)41°F to 140°F, 39.2°F to 140°F
Temperature Resolution±0.1°C
Temperature Uniformity0.2°at 37°C (in Flask)
Frequency50 Hz
Depth (English) Interior21.1 in.
Depth (Metric) Interior53.6 cm
Dimensions (D x W x H) Interior21.1 x 34.3 x 18.5 in. (53.6 x 87.1 x 47 cm)
Dimensions (L x W x H)30 x 45 x 40.4 in. (76.2 x 114.3 x 102.6 cm)
Height (English)40.4 in.
Height (English) Interior18.5 in.
Height (Metric)102.6 cm
Height (Metric) Interior47 cm
Length (English)30 in.
Length (English) Platform29.5 in.
Length (Metric)76.2 cm
Length (Metric) Platform74.9 cm
Width (English)45 in.
Width (English) Interior34.3 in.
Width (Metric)114.3 cm
Width (Metric) Interior87.1 cm
Width (Metric) Platform45.7 cm
Shipping Weight (English)665 lb.
Shipping Weight (Metric)302 kg
Weight (English)635 lb.
Weight (Metric)288 kg
Controller TypeDigital
DescriptionIncubated/Refrigerated Floor-Model Console Orbital Shaker
ModelRefrigerated and Incubated Console Shakers
Humidity Range0% at/below 31°C (87.8°F)
Motor TypeDC brushless
Timer RangeContinuous or timed operation from 1 minute up to 199 hoursand 59 minutes
Unit SizeEA