Thermolyne™ Premium Large Muffle Furnaces Accessories
Product Image
Thermolyne™ Premium Large Muffle Furnaces Accessories
Use these accessories with Thermolyne Premium Large Muffle Furnaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SH408X1
also known as AVL
Stainless-steel Shelf
JSX16Shelf Pegs
PHX1
also known as AVL
Hearth Tray, 8.2 x 10.1 x 1.27 cm
Catalog number SH408X1
also known as AVL
Price (SGD)
176.00
EA
Add to cart
Description:
Stainless-steel Shelf
Price (SGD)
176.00
EA
Add to cart
Use Thermo Scientific™ Thermolyne Premium Large Muffle Furnace Accessories to increase the utility of the Large Muffle Furnace.
Specifications
DescriptionStainless-steel Shelf
For Use With (Equipment)Thermolyne Premium Large Muffle Furnace
TypeShelf
Length (English)8.3 in.
Width (English)12.8 in.
Unit SizeEA