Square-Bottom Antistatic Cleanroom Bags
Square-Bottom Antistatic Cleanroom Bags
Actual product may vary
Square-Bottom Antistatic Cleanroom Bags
Square-Bottom Antistatic Cleanroom Bags
Square-Bottom Antistatic Cleanroom Bags
Square-bottom, antistatic bags are treated with an amine-free agent
Have Questions?
Catalog NumberDimensions (L x W)Height (English)
SCN0205-0050472 x 32 in.80 in.
Catalog number SCN0205-00504
Price (SGD)
-
Dimensions (L x W):
72 x 32 in.
Height (English):
80 in.
Constructed from proven low-density polyethylene film. Perform well in a broad spectrum of applications. Ideal for parts that require a moderate level (Levels II and III) of packaging cleanliness.
  • Tough yet flexible
  • Chemically resistant to solvents
  • Exhibit low water vapor transmission rate and good electrical properties
Specifications
AutoclavableNot Autoclavable
ColorClear
DisposableSingle-use
MaterialPolyethylene
Dimensions (L x W)72 x 32 in.
Length (English)72 in.
Length (Metric)182.88 cm
Width (English)32 in.
Width (Metric)81.28 cm
Height (English)80 in.
Product TypeCleanroom Bags
Unit SizeEA