Square-Bottom Antistatic Cleanroom Bags
Square-Bottom Antistatic Cleanroom Bags
Actual product may vary
Square-Bottom Antistatic Cleanroom Bags
Square-Bottom Antistatic Cleanroom Bags
Square-Bottom Antistatic Cleanroom Bags
Square-bottom, antistatic bags are treated with an amine-free agent
Have Questions?
Catalog NumberDimensions (L x W)Height (English)Quantity
SCN0205-0036724 x 48 in.48 in.1 Ea.
Catalog number SCN0205-00367
Price (SGD)
19.00
EA
Add to cart
Dimensions (L x W):
24 x 48 in.
Height (English):
48 in.
Quantity:
1 Ea.
Price (SGD)
19.00
EA
Add to cart
Constructed from proven low-density polyethylene film. Perform well in a broad spectrum of applications. Ideal for parts that require a moderate level (Levels II and III) of packaging cleanliness.
  • Tough yet flexible
  • Chemically resistant to solvents
  • Exhibit low water vapor transmission rate and good electrical properties
Specifications
AutoclavableNot Autoclavable
ColorClear
DisposableSingle-use
MaterialPolyethylene
Quantity1 Ea.
Dimensions (L x W)24 x 48 in.
Length (English)24 in.
Length (Metric)60.96 cm
Width (English)48 in.
Width (Metric)121.9 cm
Height (English)48 in.
Product TypeCleanroom Bags
Unit SizeEA