Sunflower Seed Oil, Spectrum™ Chemical
Sunflower Seed Oil, Spectrum™ Chemical
Sunflower Seed Oil, Spectrum™ Chemical
Sunflower Seed Oil, Spectrum™ Chemical
Spectrum Chemical Mfg. Corp.
Sunflower Seed Oil, Spectrum™ Chemical
S1929, 8001-21-6
Have Questions?
Catalog NumberQuantityPackaging
SCMS1929-500MLGL500 mLAmber Glass Bottle
Catalog number SCMS1929-500MLGL
Price (SGD)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
CAS8001-21-6
CAS Max %1
PackagingAmber Glass Bottle
Quantity500 mL
Saponification Value188-194
View more
Spectrum™ Chemical Sunflower Seed Oil is, as its name suggests, the oil of sunflower seeds that is extracted by the compression of the seeds. This non-volatile oil is most commonly used in food and as a frying oil. It is also used in cosmetics as an emollient. It is also mixed with diesel to run diesel combustion engines.