Peanut Oil, NF, Spectrum™ Chemical
Peanut Oil, NF, Spectrum™ Chemical
Peanut Oil, NF, Spectrum™ Chemical
Spectrum Chemical Mfg. Corp.
Peanut Oil, NF, Spectrum™ Chemical
PE090, 8002-03-7
Have Questions?
Catalog NumberQuantity
SCMPE090-500MLGL500 mL
Catalog number SCMPE090-500MLGL
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPeanut Oil
CAS8002-03-7
CAS Max %1
PackagingAmber Glass Bottle
Quantity500 mL
View more
Spectrum™ Chemical Peanut Oil, NF is used as a vehicle for pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.