Check Order Status
DURAN™ GL 45 Pouring Rings For Screw Caps
Actual product may vary
Duran
DURAN™ GL 45 Pouring Rings For Screw Caps
Distinguishing media types is simplified.
Have Questions?
Change view
Catalog Number
Color
SCH292422809
Blue
FIS09-841-240
Blue
2 Options
Catalog number
SCH292422809
Price (SGD)
18.00
Case
Add to cart
Color:
Blue
Price (SGD)
18.00
Case
Add to cart
Available in blue, green, yellow and grey
Specifications
Color
Blue
Height (Metric) With Closure
4mm
Material
Polypropylene
Product Type
Pouring Ring
For Use With (Equipment)
Screw Cap
Autoclavable
Autoclavable