Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Stoppers and Closures
▸
Bottle Caps
▸
DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles
Actual product may vary
Schott Duran
DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles
Distinguishing media types is simplified and interchanging of screw caps and carry over of substances is effectively excluded.
Have Questions?
Catalog Number
Color
Material
SCH292402807
Red
PBT
Catalog number
SCH292402807
Price (SGD)
174.00
Pack
Add to cart
Color:
Red
Material:
PBT
Price (SGD)
174.00
Pack
Add to cart
Available in blue, green, yellow and grey with match pour rings
Specifications
Color
Red
Material
PBT
Sterility
Non-sterile
Thread Size (Metric)
GL 45
For Use With (Equipment)
Laboratory glass bottles
Closure Size
GL 45
Autoclavable
Autoclavable
Disposable
Reusable
Product Type
Screw Cap