▸
Stoppers and Closures
▸
Bottle Caps
▸
DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles
Actual product may vary
Duran
DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles
Distinguishing media types is simplified and interchanging of screw caps and carry over of substances is effectively excluded.
Have Questions?
Change view
Catalog Number
Color
Material
SCH292392809
Blue
Polypropylene
SCH292402807
Red
PBT
2 Options
Catalog number
SCH292392809
Price (SGD)
41.00
Case
Add to cart
Color:
Blue
Material:
Polypropylene
Price (SGD)
41.00
Case
Add to cart
Available in blue, green, yellow and grey with match pour rings
Specifications
Color
Blue
Material
Polypropylene
Thread Size (Metric)
GL 45
Product Type
Screw Cap
For Use With (Equipment)
Laboratory glass bottles
Closure Size
GL 45