▸
Bottles, Jars and Jugs
▸
Bottles
▸
Media Bottles
▸
DURAN™ GL 45 Amber Laboratory Glass Bottles w/o Screw-cap and Pour Ring
Actual product may vary
Duran
DURAN™ GL 45 Amber Laboratory Glass Bottles w/o Screw-cap and Pour Ring
UV protection up to approx. 500 nm wavelength. Unchanged DURAN™ properties within the bottle, as coloration is only to the outer surface.
Have Questions?
Catalog Number
Capacity (Metric)
SCH218064406
500 mL
Catalog number
SCH218064406
Price (SGD)
560.00
Pack
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
560.00
Pack
Add to cart
With easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic
Select sizes available with retrace code
Specifications
Capacity (Metric)
500 mL
Color
Amber
Material
Glass
Thread Size (Metric)
GL 45
Height (Metric)
176 mm
Product Type
Laboratory Bottle
Diameter (Metric)
86 mm