DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring
With proven DURAN™ properties. Available as bottle only and completed bottle with blue screw cap (PP1, integral lip seal) and pouring ring (PP1) for drip-free pouring and clean, safe working.
Have Questions?
Catalog Number
Capacity (Metric)
SCH218014459
500 mL
Catalog number SCH218014459
Price (SGD)
134.00
Case
Capacity (Metric):
500 mL
Price (SGD)
134.00
Case
Service temperature level of screw cap and pouring ring: +140 °C
Typical applications: storage, sample preparation, transport