▸
Bottles, Jars and Jugs
▸
Bottles
▸
Media Bottles
▸
DURAN™ GL 45 Laboratory Glass Bottles Without PP Screw Cap and Pouring Ring
Actual product may vary
Schott Duran
DURAN™ GL 45 Laboratory Glass Bottles Without PP Screw Cap and Pouring Ring
For general laboratory use.
Have Questions?
Catalog Number
Capacity (Metric)
SCH218014401
500 mL
Catalog number
SCH218014401
Price (SGD)
110.00
Case
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
110.00
Case
Add to cart
With DIN thread and graduation
Specifications
Capacity (Metric)
500 mL
Color
Clear
Material
Glass
Height (Metric)
176 mm
Diameter (Metric)
86 mm