DURAN™ Museum Jar, with ground glass plate
DURAN™ Museum Jar, with ground glass plate
Actual product may vary
DURAN™ Museum Jar, with ground glass plate
Museum jars are used to store materials. The large glass surfaces allow a good view into the glass on its contents.
Have Questions?
Catalog NumberDimensions (L x W x H)
SCH213634703210 x 210 x 100 mm
Catalog number SCH213634703
Price (SGD)
387.00
EA
Add to cart
Dimensions (L x W x H):
210 x 210 x 100 mm
Price (SGD)
387.00
EA
Add to cart
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Exhibition glasses and museum jars are used to store materials and are often used in exhibitions. Here the objects on display can be preserved and then viewed without touching them.
  • The precise grinding of the glass plate and base vessel enables a very tight seal
  • Surface plate
Specifications
Dimensions (L x W x H)210 x 210 x 100 mm
For Use With (Application)Exhibition of specimen; Storage
MaterialBorosilicate Glass (3.3)
Quantity1 Ea.