DURAN™ Low Form Beakers with Spout
Actual product may vary
DURAN™ Low Form Beakers with Spout
DURAN™ Low Form Beakers with Spout
Schott Duran
DURAN™ Low Form Beakers with Spout
Uniform wall thickness distribution makes these beakers ideal for heating applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SCH211064103400 mL
FIS09-841-1041 L
SCH2110654081 L
SCH2110663012000 mL
FIS09-841-1065 L
Catalog number SCH211064103
Price (SGD)
86.00
Case
Capacity (Metric):
400 mL
Price (SGD)
86.00
Case
  • Easy-to-read scale and large labelling field for easy marking in fired-on, highly durable, white ceramic
  • Spout for clean pouring
Specifications
Capacity (Metric)400 mL
MaterialBorosilicate Glass
Height (Metric)110 mm
Quantity10/Pk.
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
ScaleWhite Scale
StyleGriffin, Low Form
Unit SizeCase