Check Order Status
Polypropylene Membrane Screw Caps
Actual product may vary
Polypropylene Membrane Screw Caps
Have Questions?
Catalog Number
ThreadSizeMetric
SCH1183251
Catalog number
SCH1183251
Price (SGD)
-
Specifications
Color
Blue
Material
Polypropylene
Quantity
5Pack
Height (Metric)
24 mm
Product Type
Membrane Screw Cap
For Use With (Equipment)
Pressure Equalization
Closure Size
GL 32
Diameter (Metric)
41 mm