▸
Evaporators
▸
Evaporator Accessories
▸
SCC1 CryoCool Heat Transfer Fluid
SCC1 CryoCool Heat Transfer Fluid
Savant™ SCC1 CryoCool Heat Transfer Fluid is suitable for use with the Explorer SpeedVac Vacuum Concentrator.
Have Questions?
Catalog Number
For Use With (Equipment)
SCC1
also known as AVL
Explorer SpeedVac Vacuum Concentrator
Catalog number
SCC1
also known as AVL
HSN
38249090
Price (SGD)
313.00
Each
Add to cart
For Use With (Equipment):
Explorer SpeedVac Vacuum Concentrator
Price (SGD)
313.00
Each
Add to cart
Suitable for use with Explorer SpeedVac Vacuum Concentrator
Specifications
Description
CryoCool™ Heat Transfer Fluid, 1L
Type
Heat Transfer Fluid
For Use With (Equipment)
Explorer SpeedVac Vacuum Concentrator
Model
SCC1
Volume (Metric)
1 L