Revco™ High-Performance Laboratory Refrigerators with Solid Doors
Revco™ High-Performance Laboratory Refrigerators with Solid Doors
Actual product may vary
Revco™ High-Performance Laboratory Refrigerators with Solid Doors
Store biological and chemical samples for clinical, industrial and scientific applications in these high-performance laboratory refrigerators with solid doors.
Have Questions?
Catalog NumberCapacity (English)VoltagePlug Type
REL2304V
also known as AVL
23.3 cu. ft.230 VEU
Catalog number REL2304V
also known as AVL
Price (SGD)
-
Capacity (English):
23.3 cu. ft.
Voltage:
230 V
Plug Type:
EU
Store samples for clinical, industrial and scientific applications in Thermo Scientific™ Revco™ High-Performance Laboratory Refrigerators with Solid Doors. These refrigerators feature a microprocessor control system, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors.
Key Features
  • Adjustable temperatures between 1° and 8°C (factory preset at 4°C)
  • Automatic defrost and condensate removal
Microprocessor Control
  • Digital electronic technology designed to simplify operation and deliver greater accuracy
  • Icon-based controls
  • Large, easy-to-read digital display of temperature within 0.1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for over temperature and flashes bulb for under temperature
  • Sensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost build-up and optimizing coil efficiency
Alarm System
  • Audible/visual warnings for temperature deviations or power failure
  • Key-operated ON/OFF/ALARM ON master switch for setpoint security
  • Push-button alarm test and remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system
  • Alarm setpoints are preset to warn of temperature deviation at 5.5°C over temperature and 1.5°C under temperature
Construction
  • Positive forced-air circulation systems
  • Industrial-grade cabinet construction and compressors
  • High-density CFC-free urethane foam insulation
  • Self-closing, key-locked doors
  • Epoxy-coated, open-wire shelves adjustable in 1 in. (2.5 cm) increments
  • Cold-rolled steel interior and exterior finished with high-impact powder paint coating
  • Interior lights with door-activated ON/OFF switch and independent switch on control panel
  • Adjustable leveling feet (except 4.9 cu. ft. undercounter model)
  • 45.8 cu. ft. (1297 L) units have double sliding glass doors, base-mounted controls and refrigeration systems, independent switch-activated lights, and dual-wheel 2 in. (5 cm) casters instead of leveling feet
  • Undercounter model has a 1 in. (2.5 cm) dia. access port with plug
Ordering Information:
Alarm systems, temperature chart recorders, and power monitors are available separately. Other voltages are available; contact your Customer Service or Sales Representative for more information.
Warranty varies by country, please contact us for details
Specifications
TypeHigh-Performance Refrigerator
Capacity (English)23.3 cu. ft.
Capacity (Metric)659 L
Temperature Range1°C to 8°C
RefrigerantCFC-free
Door StyleSolid
No. of Doors1
Shelves4
Cabinet MaterialCold-rolled Steel
Exterior FinishHigh-impact Powder Paint Coating
InsulationCFC-free
Certifications/ComplianceCE, cUL, TUV, UL
StyleUpright
Casters2 in. (5cm) casters (two locking, two nonlocking), factory installed
Control TypeMicroprocessor
DefrostAutomatic
Monitoring OptionsAlarm, Recorder
CompressorIndustrial-grade
Dimensions (D x W x H) Interior29 x 24 x 58 in. (73.7 x 61 x 147.3 cm)
Dimensions (L x W x H) Exterior37.2 x 28 x 79.2 in. (94.5 x 71 x 201.2 cm)
Depth (English) Interior29 in.
Depth (Metric) Interior73.7 cm
Height (English) Exterior79.2 in.
Height (English) Interior58 in.
Height (Metric) Exterior201.2 cm
Height (Metric) Interior147.3 cm
Length (English) Exterior37.2 in.
Length (Metric) Exterior94.5 cm
Width (English) Exterior28 in.
Width (English) Interior24 in.
Width (Metric) Exterior71 cm
Width (Metric) Interior61 cm
Amperage6 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Shipping Weight (English)425 lb.
Shipping Weight (Metric)193 kg
Unit SizeEA