Product Image
Thermo Scientific Rotor Block
Thermo Scientific Rotor Block, Aluminium
Have Questions?
Catalog NumberCapacity
RBA24-17-6024 Tubes
Catalog number RBA24-17-60
HSN 84219100
Price (SGD)
1,452.00
Special offer
Ends: 30-Jun-2026
1,815.00
Save 363.00 (20%)
Each
Capacity:
24 Tubes
Price (SGD)
1,452.00
Special offer
Ends: 30-Jun-2026
1,815.00
Save 363.00 (20%)
Each
Thermo Scientific Rotor Block, Aluminium, holds 24 17 x 60mm tubes, set of 2
  • Aluminium rotor block.
  • Holds 24 tubes.
Specifications
MaterialAluminum
TypeRotor
Capacity24 Tubes
For Use With (Equipment)17 x 60 mm Tubes
Unit SizeEach